Merck
  • The link between ankylosing spondylitis, Crohn's disease, Klebsiella, and starch consumption.
The link between ankylosing spondylitis, Crohn's disease, Klebsiella, and starch consumption.
Clinical & developmental immunology (2013-06-20)
Taha Rashid, Clyde Wilson, Alan Ebringer
ABSTRACT
Both ankylosing spondylitis (AS) and Crohn's disease (CD) are chronic and potentially disabling interrelated conditions, which have been included under the group of spondyloarthropathies. The results of a large number of studies support the idea that an enteropathic pathogen, Klebsiella pneumoniae, is the most likely triggering factor involved in the initiation and development of these diseases. Increased starch consumptions by genetically susceptible individuals such as those possessing HLA-B27 allelotypes could trigger the disease in both AS and CD by enhancing the growth and perpetuation of the Klebsiella microbes in the bowel. Exposure to increased levels of these microbes will lead to the production of elevated levels of anti-Klebsiella antibodies as well as autoantibodies against cross-reactive self-antigens with resultant pathological lesions in the bowel and joints. Hence, a decrease of starch-containing products in the daily dietary intake could have a beneficial therapeutic effect on the disease especially when used in conjunction with the currently available medical therapies in the treatment of patients with AS and CD.
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from rice
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10