Merck
  • Solid-phase extraction in the determination of gold, palladium, and platinum.
Solid-phase extraction in the determination of gold, palladium, and platinum.
Journal of separation science (2012-06-27)
Elisaveta Mladenova, Irina Karadjova, Dimiter L Tsalev
ABSTRACT
A simple classification of various sorbents and solid-phase extraction procedures used for preconcentration of trace levels of Au, Pd, and Pt from different sample types is proposed in this review article. The large variety of available sorbents/procedures has been organized according to expected mechanisms of sorption process (complex formation; ion exchange; adsorption; ion-imprinted or molecularly imprinted polymers); according to the kind of monomeric units of the polymer matrix as well as on the basis of the kind of functional group responsible for main performance characteristics (selectivity, capacity) of the sorbent. Advantages of chemically modified sorbents, sulfur-containing sorbent extractants, and ion-imprinted polymers, together with rational pretreatment by means of microwave treatments, scaling down of enrichment, and quantification by means of flow and flow injection approaches are given. Preferred instrumental techniques for quantification of ppb levels of Au, Pd, and Pt in prepared concentrates/column eluates are multielement instrumental techniques: inductively coupled plasma optical emission spectrometry (ICP-OES), and inductively coupled plasma mass spectrometry (ICPMS). Excellent limits of detection at picogram levels of these analytes are provided by electrothermal atomic absorption spectrometry (ETAAS), generally in single-element mode and the neutron activation analysis (NAA), while X-ray fluorescence spectrometry and flame AAS are rarely applied because of lack of sensitivity at sub-ppm levels of Au, Pd, and Pt. Some problems of atomic spectrometric quantification techniques and their representative limits of detection are given. Recent applications to geological, industrial, pharmaceutical, biological, and other materials are tabulated. References have been selected mostly from the period 1995 to 2010.
Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 6mm disks, thickness 0.025mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 0.2m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 10mm disks, thickness 0.0125mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, nanopowder, 200 nm particle size (SEM), 99.9% (metals basis)
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 0.2m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 10mm disks, thickness 0.0075mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 10m, diameter 0.08mm, hard, 99.9%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 6mm disks, thickness 0.005mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.998%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 0.05m, diameter 0.175mm, as drawn, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 0.1m, diameter 0.15mm, annealed, may be used as one component of a thermocouple, 99.99%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 8mm disks, thickness 0.005mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, foil, 4mm disks, thickness 0.01mm, as rolled, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Platinum, wire reel, 0.1m, diameter 0.006mm, wollaston, 99.9%