Merck
  • Interactions between nitric oxide and plant hormones in aluminum tolerance.
Interactions between nitric oxide and plant hormones in aluminum tolerance.
Plant signaling & behavior (2012-04-14)
Huyi He, Longfei He, Minghua Gu
ABSTRACT
Nitric oxide (NO) is involved, together with plant hormones, in the adaptation to Al stress in plants. However, the mechanism by which NO and plant hormones interplay to improve Al tolerance are still unclear. We have recently shown that patterns of plant hormones alteration differ between rye and wheat under Al stress. NO may enhance Al tolerance by regulating hormonal equilibrium in plants, as a regulator of plant hormones signaling. In this paper, some unsolved issues are discussed based on recent studies and the complex network of NO and plant hormones in inducing Al tolerance of plants are proposed.
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, sphere, 100pcs, diameter 1.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, wire reel, 1000m, diameter 0.30mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, sphere, 100pcs, diameter 2.5mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, rod, 200mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, microleaf disks, 25mm, thinness 0.15μm, specific density 41.28μg/cm2, removable support, 99.999%
Aluminum, mesh, 900x900mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, tube, 1000mm, outside diameter 1.42mm, inside diameter 1.166mm, wall thickness 0.127mm, as drawn, 99.5%
Aluminum, rod, 200mm, diameter 20.0mm, as drawn, 99.999%
Aluminum, sphere, 100pcs, diameter 3.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, rod, 100mm, diameter 28mm, as drawn, 99.999%
Aluminum, microfoil, disks, 25mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, insulated wire, 2m, conductor diameter 0.1mm, insulation thickness 0.005mm, polyimide insulation, 99.95+%
Aluminum, tube, 1000mm, outside diameter 0.86mm, inside diameter 0.76mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, rod, 200mm, diameter 5.0mm, as drawn, 99.999%
Aluminum, tube, 500mm, outside diameter 3.0mm, inside diameter 1.62mm, wall thickness 0.69mm, hard, 99.5%
Aluminum, tube, 200mm, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, rod, 200mm, diameter 12.0mm, as drawn, 99.999%
Aluminum, wire reel, 100m, diameter 0.25mm, as drawn, 99.5%
Aluminum, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%