Merck
Niacin: chemical forms, bioavailability, and health effects.
Nutrition reviews (2012-06-01)
Douglas MacKay, John Hathcock, Erminia Guarneri
ABSTRACT
Elevated low-density lipoprotein cholesterol (LDL-C) has been the main target of lipid-altering therapy to reduce cardiovascular risk associated with dyslipidemia. Residual cardiovascular risk remains, however, after achievement of goal LDL-C levels and is associated in part with other risk markers of cardiovascular disease, including low high-density lipoprotein cholesterol (HDL-C), high lipoprotein a, and hypertriglyceridemia. Niacin is considered a valuable agent for therapy to modify high LDL-C as well as low HDL-C, high lipoprotein a, and hypertriglyceridemia. The forms of niacin available in the marketplace include unbound niacin, or free nicotinic acid (NA); extended-release NA, a form of NA that is released gradually over a period of time; inositol hexanicotinate, six molecules of NA covalently bonded to one molecule of inositol; and nicotinamide, or niacinamide, the amide form of NA, which is readily bioavailable. This review is designed to assist healthcare professionals in evaluating the form(s) of niacin best suited for a particular therapeutic goal. Further, it provides a literature-based evaluation of risk for NA, extended-release NA, inositol hexanicotinate, and nicotinamide.
Product Description
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
USP
Niacin, United States Pharmacopeia (USP) Reference Standard
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material