Merck
  • Combination of probenecid-sulphadoxine-pyrimethamine for intermittent preventive treatment in pregnancy.
Combination of probenecid-sulphadoxine-pyrimethamine for intermittent preventive treatment in pregnancy.
Malaria journal (2012-02-11)
Julie Gutman, S Patrick Kachur, Laurence Slutsker, Alexis Nzila, Theonest Mutabingwa
ABSTRACT
The antifolate sulphadoxine-pyrimethamine (SP) has been used in the intermittent prevention of malaria in pregnancy (IPTp). SP is an ideal choice for IPTp, however, as resistance of Plasmodium falciparum to SP increases, data are accumulating that SP may no longer provide benefit in areas of high-level resistance. Probenecid was initially used as an adjunctive therapy to increase the blood concentration of penicillin; it has since been used to augment concentrations of other drugs, including antifolates. The addition of probenecid has been shown to increase the treatment efficacy of SP against malaria, suggesting that the combination of probenecid plus SP may prolong the useful lifespan of SP as an effective agent for IPTp. Here, the literature on the pharmacokinetics, adverse reactions, interactions and available data on the use of these drugs in pregnancy is reviewed, and the possible utility of an SP-probenecid combination is discussed. This article concludes by calling for further research into this potentially useful combination.
Product Description
Supelco
Pyrimethamine, VETRANAL®, analytical standard
USP
Probenecid, United States Pharmacopeia (USP) Reference Standard
Pyrimethamine, European Pharmacopoeia (EP) Reference Standard
Probenecid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Probenecid