Merck
Cladribine as a therapeutic option in multiple sclerosis.
Clinical immunology (Orlando, Fla.) (2011-07-08)
Clemens Warnke, Verena I Leussink, Norbert Goebels, Orhan Aktas, Alexey Boyko, Bernd C Kieseier, Hans-Peter Hartung
ABSTRACT
There is an unmet need for more potent and convenient drugs in the treatment of patients diagnosed with multiple sclerosis (MS). Among five currently investigated oral drugs with an ongoing or completed phase III program, promising efficacy data for oral cladribine have recently been published. However, benefits need to be weighed against potential risks to define the role of this compound within current treatment regimens. Here we review present data on efficacy of oral cladribine and discuss known and anticipated risks of this drug.
Product Description
Cladribine, European Pharmacopoeia (EP) Reference Standard
Cladribine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Chloro-2′-deoxyadenosine, antileukemic