Merck
  • Aluminum stress and its role in the phospholipid signaling pathway in plants and possible biotechnological applications.
Aluminum stress and its role in the phospholipid signaling pathway in plants and possible biotechnological applications.
IUBMB life (2011-09-10)
Wilberth Poot-Poot, Soledad M Teresa Hernandez-Sotomayor
ABSTRACT
An early response of plants to environmental signals or abiotic stress suggests that the phospholipid signaling pathway plays a pivotal role in these mechanisms. The phospholipid signaling cascade is one of the main systems of cellular transduction and is related to other signal transduction mechanisms. These other mechanisms include the generation of second messengers and their interactions with various proteins, such as ion channels. This phospholipid signaling cascade is activated by changes in the environment, such as phosphate starvation, water, metals, saline stres, and plant-pathogen interactions. One important factor that impacts agricultural crops is metal-induced stress. Because aluminum has been considered to be a major toxic factor for agriculture conducted in acidic soils, many researchers have focused on understanding the mechanisms of aluminum toxicity in plants. We have contributed the last fifteen years in this field by studying the effects of aluminum on phospholipid signaling in coffee, one of the Mexico's primary crops. We have focused our research on aluminum toxicity mechanisms in Coffea arabica suspension cells as a model for developing future contributions to the biotechnological transformation of coffee crops such that they can be made resistant to aluminum toxicity. We conclude that aluminum is able to not only generate a signal cascade in plants but also modulate other signal cascades generated by other types of stress in plants. The aim of this review is to discuss possible involvement of the phospholipid signaling pathway in the aluminum toxicity response of plant cells.
Product Description
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, rod, 100mm, diameter 6.0mm, as drawn, 99.999%
Aluminum, microfoil, disks, 50mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 100mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, rod, 44mm, diameter 77.5mm, as drawn, 99.999%
Aluminum, tube, 200mm, outside diameter 0.86mm, inside diameter 0.76mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, microfoil, disks, 25mm, thinness 0.0075μm, specific density 1.84μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 0.5m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, wire reel, 100m, diameter 0.5mm, as drawn, 99.95+%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, tube, 100mm, outside diameter 0.6mm, inside diameter 0.3mm, wall thickness 0.15mm, as drawn, 99.5%
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, tube, 200mm, outside diameter 1.42mm, inside diameter 1.166mm, wall thickness 0.127mm, as drawn, 99.5%
Aluminum, rod, 200mm, diameter 10.0mm, as drawn, 99.999%
Aluminum, tube, 500mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, tube, 500mm, outside diameter 3.0mm, inside diameter 1.62mm, wall thickness 0.69mm, hard, 99.5%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, sphere, 100pcs, diameter 1.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, wire reel, 100m, diameter 0.025mm, as drawn, 99.0+%
Aluminum, rod, 200mm, diameter 12.7mm, 99%
Aluminum, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%