Merck
Degradable polyethylene: fantasy or reality.
Environmental science & technology (2011-04-19)
Prasun K Roy, Minna Hakkarainen, Indra K Varma, Ann-Christine Albertsson
ABSTRACT
Plastic waste disposal is one of the serious environmental issues being tackled by our society today. Polyethylene, particularly in packaging films, has received criticism as it tends to accumulate over a period of time, leaving behind an undesirable visual footprint. Degradable polyethylene, which would enter the eco-cycle harmlessly through biodegradation would be a desirable solution to this problem. However, the "degradable polyethylene" which is presently being promoted as an environmentally friendly alternative to the nondegradable counterpart, does not seem to meet this criterion. This article reviews the state of the art on the aspect of degradability of polyethylene containing pro-oxidants, and more importantly the effect these polymers could have on the environment in the long run. On exposure to heat, light, and oxygen, these polymers disintegrate into small fragments, thereby reducing or increasing the visual presence. However, these fragments can remain in the environment for prolonged time periods. This article also outlines important questions, particularly in terms of time scale of complete degradation, environmental fate of the polymer residues, and possible accumulation of toxins, the answers to which need to be established prior to accepting these polymers as environmentally benign alternatives to their nondegradable equivalents. It appears from the existing literature that our search for biodegradable polyethylene has not yet been realized.
Product Description
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Polyethylene (LDPE), ERM®, certified reference material