Merck
Acetazolamide and sulfonamide allergy: a not so simple story.
High altitude medicine & biology (2010-12-31)
Thomas E Kelly, Peter H Hackett
ABSTRACT
Allergies and adverse reactions to sulfonamide medications are quite common. Two distinct categories of drugs are classified as sulfonamides: antibiotics and nonantibiotics. The two groups differ in their chemical structure, use, and the rate at which adverse reactions occur. Cross-reactivity between the two groups has been implied in the past, but is suspect. Acetazolamide, from the nonantibiotic group, is routinely used in the prevention and treatment of high altitude issues and may not need to be avoided in individuals with a history of sulfonamide allergy. This review addresses the differences between the groups and the propensity for intergroup and intragroup adverse reactions based on the available literature. We also examine the different clinical presentations of allergy and adverse reactions, from simple cutaneous reactions with no sequelae through Stevens-Johnson syndrome and anaphylaxis, with risk for significant morbidity and mortality. We offer a systematic approach to determine whether acetazolamide is a safe option for those with a history of allergy to sulfonamides.
Sigma-Aldrich
Acetazolamide, ≥99%, powder
Acetazolamide, European Pharmacopoeia (EP) Reference Standard
Acetazolamide for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Acetazolamide, United States Pharmacopeia (USP) Reference Standard