Merck
Oleoyl-estrone.
Medicinal research reviews (2011-02-03)
Xavier Remesar, José Antonio Fernández-López, Marià Alemany
ABSTRACT
Oleoyl-estrone (OE) is a powerful slimming agent that is also present in plasma and adipose tissue, where it is synthesized. It acts through the formation of a derivative W. OE effects (and W levels) are proportional to the dose. OE reduces food intake but maintains energy expenditure (thermogenesis). The energy gap is fulfilled with adipose tissue fat, sparing body protein and maintaining glycemia (and glycogen) with lower insulin and leptin levels. OE (in fact W) acts through specific receptors, different from those of estrogen. OE increases cholesterol catabolism, reducing hypercholesterolemia in obese rats. The main metabolic effect on adipose tissue is lowering of lipid synthesis, maintaining unchanged the intracellular lipolytic processes; the imbalance favors the progressive loss of fat, which is largely used by the muscle. OE administration induces additive effects with other antiobesity agents, such as β(3)-adrenergic agonists, forcing a massive loss of lipid. Corticosteroids markedly limit OE action by altering the liver control of lipogenesis. OE also inhibits the action of 17β-hydroxysteroid dehydrogenase, decreasing the synthesis of β-estradiol and testosterone. Discontinuous treatment allows for maximal efficacy both in rats and humans. OE has the advantage that the loss of fat is maintained and does not require additional dietary limitations.
Product Description
Supelco
Estrone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Estrone, European Pharmacopoeia (EP) Reference Standard
USP
Estrone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estrone, ≥99%
Supelco
Estrone, Pharmaceutical Secondary Standard; Certified Reference Material