Merck
  • Pyridoxal-5'-phosphate-dependent enzymes involved in biotin biosynthesis: structure, reaction mechanism and inhibition.
Pyridoxal-5'-phosphate-dependent enzymes involved in biotin biosynthesis: structure, reaction mechanism and inhibition.
Biochimica et biophysica acta (2010-12-25)
Stéphane Mann, Olivier Ploux
ABSTRACT
The four last steps of biotin biosynthesis, starting from pimeloyl-CoA, are conserved among all the biotin-producing microorganisms. Two enzymes of this pathway, the 8-amino-7-oxononanoate synthase (AONS) and the 7,8-diaminopelargonic acid aminotransferase (DAPA AT) are dependent on pyridoxal-5'-phosphate (PLP). This review summarizes our current understanding of the structure, reaction mechanism and inhibition on these two interesting enzymes. Mechanistic studies as well as the determination of the crystal structure of AONS have revealed a complex mechanism involving an acylation with inversion of configuration and a decarboxylation with retention of configuration. This reaction mechanism is shared by the homologous 5-aminolevulinate synthase and serine palmitoyltransferase. While the reaction catalyzed by DAPA AT is a classical PLP-dependent transamination, the inactivation of this enzyme by amiclenomycin, a natural antibiotic that is active against Mycobacterium tuberculosis, involves the irreversible formation of an adduct between PLP and amiclenomycin. Mechanistic and structural studies allowed the complete description of this unique inactivation mechanism. Several potent inhibitors of these two PLP-dependent enzymes have been prepared and might be useful as starting points for the design of herbicides or antibiotics. This article is part of a Special Issue entitled: Pyridoxal Phospate Enzymology.
Product Description
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Biotin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
USP
Biotin, United States Pharmacopeia (USP) Reference Standard
SAFC
BIOTIN
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material