Merck
Sodium bicarbonate plus N-acetylcysteine prophylaxis: a meta-analysis.
JACC. Cardiovascular interventions (2009-11-21)
Jeremiah R Brown, Clay A Block, David J Malenka, Gerald T O'Connor, Anton C Schoolwerth, Craig A Thompson
ABSTRACT
We sought to conduct a meta-analysis to compare N-acetylcysteine (NAC) in combination with sodium bicarbonate (NaHCO(3)) for the prevention of contrast-induced acute kidney injury (AKI). Contrast-induced AKI is a serious consequence of cardiac catheterizations and percutaneous coronary interventions (PCI). Despite recent supporting evidence for combination therapy, not enough has been done to prevent the occurrence of contrast-induced AKI prophylactically. Published randomized controlled trial data were collected from OVID/PubMed, Web of Science, and conference abstracts. The outcome of interest was contrast-induced AKI, defined as a >or=25% or >or=0.5 mg/dl increase in serum creatinine from baseline. Secondary outcome was renal failure requiring dialysis. Ten randomized controlled trials met our criteria. Combination treatment of NAC with intravenous NaHCO(3) reduced contrast-induced AKI by 35% (relative risk: 0.65; 95% confidence interval: 0.40 to 1.05). However, the combination of N-acetylcysteine plus NaHCO(3) did not significantly reduce renal failure requiring dialysis (relative risk: 0.47; 95% confidence interval: 0.16 to 1.41). Combination prophylaxis with NAC and NaHCO(3) substantially reduced the occurrence of contrast-induced AKI overall but not dialysis-dependent renal failure. Combination prophylaxis should be incorporated for all high-risk patients (emergent cases or patients with chronic kidney disease) and should be strongly considered for all interventional radio-contrast procedures.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C