Merck
Fluconazole prophylaxis in high-risk neonates.
The Annals of pharmacotherapy (2009-12-31)
Brent N Reed, Kelly E Caudle, P David Rogers
ABSTRACT
To evaluate the literature regarding the use of fluconazole prophylaxis in high-risk neonates. Literature was accessed through MEDLINE (February 2001-August 2009) using the search terms fluconazole and prophylaxis, with limits for age group (ie, birth-18 y). Reference citations from identified articles were also reviewed. DATA SELECTION AND DATA EXTRACTION: All prospective and retrospective studies in English identified from MEDLINE were evaluated. Critically ill neonates possess a number of risk factors that predispose them to fungal colonization with Candida spp. In many cases, colonization may progress to invasive systemic infections despite efforts aimed at early diagnosis and treatment. Because of its success among immunocompromised patients, fluconazole prophylaxis has been suggested as a possible approach for reducing the rates of both colonization and invasive fungal infections among at-risk neonates. To date, 4 prospective randomized controlled trials and 8 retrospective cohort studies have examined fluconazole prophylaxis in neonates. Although fluconazole prophylaxis appears to reduce the rates of colonization and invasive fungal infections, no trial in this review was able to demonstrate a significant difference in long-term morbidity or mortality. Concerns also remain regarding the adverse effects associated with prolonged exposure to fluconazole therapy. Lack of standardized study designs and treatment regimens also limit widespread recommendation for the use of fluconazole prophylaxis in clinical practice. While it may be beneficial for critically ill neonates with certain predisposing risk factors (eg, central venous access, sustained exposure to broad-spectrum antibiotics, or units with significantly high incidence of invasive fungal infections), existing research does not support the use of fluconazole prophylaxis based on birth weight or gestational age alone. Multifactor analysis evaluating the effect of fluconazole prophylaxis is necessary to establish which neonates would benefit from this practice.
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluconazole solution, 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®