Merck
  • Macrolides, ketolides, and glycylcyclines: azithromycin, clarithromycin, telithromycin, tigecycline.
Macrolides, ketolides, and glycylcyclines: azithromycin, clarithromycin, telithromycin, tigecycline.
Infectious disease clinics of North America (2009-11-17)
Jerry M Zuckerman, Fozia Qamar, Bartholomew R Bono
ABSTRACT
The advanced macrolides, azithromycin and clarithromycin, and the ketolide, telithromycin, are structural analogs of erythromycin. They have several distinct advantages when compared with erythromycin, including enhanced spectrum of activity, more favorable pharmacokinetics and pharmacodynamics, once-daily administration, and improved tolerability. Clarithromycin and azithromycin are used extensively for the treatment of respiratory tract infections, sexually transmitted diseases, and Helicobacter pylori-associated peptic ulcer disease. Telithromycin is approved for the treatment of community-acquired pneumonia. Severe hepatotoxicity has been reported with the use of telithromycin.
Product Description
USP
Clarithromycin Identity, United States Pharmacopeia (USP) Reference Standard
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard