Merck
Metallosis after metal-on-polyethylene total hip arthroplasty.
American journal of orthopedics (Belle Mead, N.J.) (2008-04-11)
Cara A Cipriano, Paul S Issack, Burak Beksac, Alejandro Gonzalez Della Valle, Thomas P Sculco, Eduardo A Salvati
ABSTRACT
Metal debris should not be generated in a well-fixed, well-functioning metal-on-polyethylene total hip arthroplasty. However, surgeons sometimes encounter periprosthetic metallosis during revision hip surgery. Insert wear, fracture, or dislodgment in modular components may lead to articulation of the prosthetic head with the metallic shell and subsequent metallosis. Metallosis may occur with loose acetabular components as a consequence of fretting of the screws and shell screw holes or shedding of the ingrowth surface of the component. The femoral component can also be a source of metallosis: Wear of a titanium femoral head, loosening of rough surface finish from the femoral stem, and stem fracture all may result in metallic particles being deposited in periarticular tissues. Specific clinical and radiographic findings can help in differentiating these forms of failure and in planning surgery. When metallic debris-induced bone loss is recognized early, surgical intervention may limit its progression.
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Supelco
Polyethylene, analytical standard, for GPC, 2,000
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, Medium density
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Polyethylene (LDPE), ERM®, certified reference material