Merck
  • Postnatal hydrocortisone treatment for chronic lung disease in the preterm newborn and long-term neurodevelopmental follow-up.
Postnatal hydrocortisone treatment for chronic lung disease in the preterm newborn and long-term neurodevelopmental follow-up.
Archives of disease in childhood. Fetal and neonatal edition (2007-09-13)
K J Rademaker, L S de Vries, C S P M Uiterwaal, F Groenendaal, D E Grobbee, F van Bel
ABSTRACT
The benefits versus the risks of postnatal administration of steroids in preterm-born infants are still debatable. This review examines the literature on postnatal hydrocortisone treatment for chronic lung disease (CLD) in preterm-born infants with a particular focus on the effects of such treatment on long-term neurodevelopmental outcomes. Quantitative published evidence does not point to a clear advantage of treatment with hydrocortisone over dexamethasone with regard to the impact on long-term neurological outcomes. However, in the absence of a randomised comparison, a consensus may soon have to be reached on the basis of the best available evidence whether hydrocortisone should replace dexamethasone in the treatment of CLD.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard