Merck
  • Drospirenone/ethinylestradiol 3mg/20microg (24/4 day regimen): a review of its use in contraception, premenstrual dysphoric disorder and moderate acne vulgaris.
Drospirenone/ethinylestradiol 3mg/20microg (24/4 day regimen): a review of its use in contraception, premenstrual dysphoric disorder and moderate acne vulgaris.
Drugs (2007-08-09)
Caroline Fenton, Keri Wellington, Marit D Moen, Dean M Robinson
ABSTRACT
Drospirenone 3mg with ethinylestradiol 20microg (Yaz) is a low-dose combined oral contraceptive (COC) administered in a regimen of 24 days of active tablets followed by a short hormone-free interval (4 days; 24/4 regimen). Drospirenone, unlike other synthetic progestogens used in COCs, is a 17alpha-spirolactone derivative and a 17alpha-spironolactone analogue with antimineralocorticoid and antiandrogenic properties. Drospirenone/ethinylestradiol 3mg/20microg (24/4) is approved in the US for the prevention of pregnancy in women, for the treatment of the symptoms of premenstrual dysphoric disorder (PMDD) and for the treatment of moderate acne vulgaris in women who wish to use an oral contraceptive for contraception.Drospirenone/ethinylestradiol 3mg/20microg (24/4) provided 99% contraceptive protection over 1 year of treatment in two large studies. The same treatment regimen over three treatment cycles also significantly improved the emotional and physical symptoms associated with PMDD, and improved moderate acne vulgaris over six treatment cycles in double-blind trials. It was generally well tolerated, with adverse events generally typical of those experienced with other COCs and which were most likely to occur in the first few cycles. Clinical trials indicate that drospirenone/ethinylestradiol 3mg/20microg (24/4) is a good long-term contraceptive option, and additionally offers relief of symptoms that characterise PMDD and has a favourable effect on moderate acne vulgaris.
Product Description
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Drospirenone, European Pharmacopoeia (EP) Reference Standard
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Drospirenone, ≥98% (HPLC)
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard