Merck
  • A review of the efficacy of fixed-dose combinations olmesartan medoxomil/hydrochlorothiazide and amlodipine besylate/benazepril in factorial design studies.
A review of the efficacy of fixed-dose combinations olmesartan medoxomil/hydrochlorothiazide and amlodipine besylate/benazepril in factorial design studies.
American journal of cardiovascular drugs : drugs, devices, and other interventions (2006-03-25)
Allen Quan, Kathleen Chavanu, Jennifer Merkel
ABSTRACT
In order to adequately control hypertension, the majority of patients will require treatment with more than one antihypertensive agent. Fixed-dose combination therapy offers several advantages, including improved efficacy, tolerability, and treatment compliance. Certain combinations have benefits in specific patient populations, such as the elderly or those with comorbidities. In this review, we evaluate the BP-lowering efficacy of olmesartan medoxomil/hydrochlorothiazide (HCTZ) and amlodipine besylate/benazepril in similarly designed, randomized, placebo-controlled studies in similar patient populations. This indirect comparison showed that both combinations significantly improve both systolic and diastolic BP compared with monotherapy with the individual agents or placebo; it also demonstrated that the combinations were well tolerated. Both combination therapies significantly improved response rates, but olmesartan medoxomil/HCTZ achieved the highest control rates compared with the individual agents. On the basis of an indirect comparison of published factorial design studies, olmesartan medoxomil/HCTZ appears to be at least as effective as amlodipine besylate/benazepril and may provide quantitatively greater reductions in diastolic BP at commonly used dosages. A randomized clinical trial comparing the two combinations is needed to confirm these findings.
Sigma-Aldrich
Amlodipine besylate, ≥98% (HPLC)
Amlodipine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Amlodipine besylate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Amlodipine besylate, United States Pharmacopeia (USP) Reference Standard
Amlodipine besylate, European Pharmacopoeia (EP) Reference Standard