Merck
  • [How I treat...topical treatment of plaque type psoriasis by the combination of calcipotriol and betamethasone dipropionate].
[How I treat...topical treatment of plaque type psoriasis by the combination of calcipotriol and betamethasone dipropionate].
Revue medicale de Liege (2006-05-10)
F Henry, C Flagothier, C Piérard-Franchimont, G E Piérard
ABSTRACT
The combination of calcipotriol and betamethasone dipropionate in a single topical formulation is a recent advance in treatment of plaque type psoriasis. Studies rooted on evidence-based medicine have been recently published. We report a synthesis and a critical analysis, critical analysis of these investigations, and translate their data with a view to apply them in daily practice.
Product Description
Supelco
Betamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Betamethasone, ≥98%
Sigma-Aldrich
Betamethasone 17,21-dipropionate
USP
Betamethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Betamethasone Dipropionate, Pharmaceutical Secondary Standard; Certified Reference Material
Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Betamethasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard