Merck
  • Inhaled tobramycin (TOBI): a review of its use in the management of Pseudomonas aeruginosa infections in patients with cystic fibrosis.
Inhaled tobramycin (TOBI): a review of its use in the management of Pseudomonas aeruginosa infections in patients with cystic fibrosis.
Drugs (2003-11-12)
Susan M Cheer, John Waugh, Stuart Noble
ABSTRACT
Specifically formulated for nebulisation using the PARI LC PLUS reusable nebuliser, tobramycin solution for inhalation (TSI) [TOBI] provides a high dose of tobramycin (an aminoglycoside antibacterial with good activity against Pseudomonas aeruginosa) to the lungs of patients with cystic fibrosis, while maintaining low serum concentrations of the drug, thus reducing the risk of systemic toxicity. Intermittent (28-day on/28-day off) treatment with TSI 300 mg twice daily significantly (p < 0.001) improved lung function and sputum P. aeruginosa density compared with placebo (randomised double-blind trials), and was significantly (p = 0.008) more effective than colistin for improvement in forced expiratory volume in 1 second (small nonblind trial) in patients aged > or =6 years with cystic fibrosis and chronic P. aeruginosa infection. Improvements in lung function were most marked in adolescent patients (aged 13-17 years) in placebo-controlled trials. Improvements were maintained for up to 96 weeks in patients in an open-label extension study. Fewer TSI than placebo recipients required parenteral antipseudomonal agents or hospitalisation. In addition, TSI 300 mg twice daily for 28 days reduced P. aeruginosa density in the lower airways of patients aged <6 years with early colonisation and cystic fibrosis, although TSI is not currently indicated in this patient group. A decrease in tobramycin susceptibility of P. aeruginosa isolates and an increase in fungal organisms (Candida albicans and Aspergillus species) during prolonged intermittent treatment with TSI 300 mg twice daily was not associated with adverse clinical outcome. There was no evidence of selection for the most resistant isolates. TSI is generally well tolerated, with no renal toxicity or hearing loss in clinical trials, although transient mild or moderate tinnitus occurred more frequently in TSI than placebo recipients. Bronchospasm after administration of TSI was transient and occurred with a similar incidence to that with placebo; TSI is preservative free and specifically formulated for the lung in terms of osmolality and pH. In conclusion, TSI provides an effective means of delivering tobramycin to the lungs of patients with cystic fibrosis with chronic P. aeruginosa infection, improving lung function and sputum P. aeruginosa density in these patients without the nephrotoxicity or ototoxicity of parenteral aminoglycosides. Further data on the potential for and clinical significance of increased tobramycin resistance and fungal colonisation during TSI treatment would be beneficial, as would longer-term data. In the meantime, TSI represents a valuable option for suppressive antipseudomonal therapy in patients with cystic fibrosis.
Product Description
Tobramycin for identification, European Pharmacopoeia (EP) Reference Standard
Tobramycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material