Merck
  • [6-endo-trig mode cyclization to a hydrindanone using samarium (II) iodide].
[6-endo-trig mode cyclization to a hydrindanone using samarium (II) iodide].
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan (2003-08-23)
Masakazu Sono
ABSTRACT
Samarium (II) iodide has been employed to promote the vinylogous pinacol coupling reaction of aldehyde to alpha, beta-unsaturated ketones. The diastereoselectivity of 6-endo-trig mode products was changed by the addition of a proton source and/or HMPA and by the reaction temperature. The stereochemistry of the hydrindanone was controlled by the coordinated samarium species, resulting in the cis-orientation in respect of the hydroxyl group at C-4 and the juncture proton at C-3a under mild reaction conditions. Coronafacic acid has been synthesized from a hydrindanone prepared by the cyclization reaction of the enone-aldehyde with samarium (II) iodide.
Sigma-Aldrich
Samarium, −40 mesh, 99% trace rare earth metals basis
Samarium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Samarium, rod, 100mm, diameter 6.35mm, cast, 99%
Samarium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Samarium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Samarium, rod, 50mm, diameter 6.35mm, cast, 99%