Merck
  • Effect of irrigation with Tetraclean on bacterial leakage of obturated root canals.
Effect of irrigation with Tetraclean on bacterial leakage of obturated root canals.
The New York state dental journal (2014-06-18)
Zahed Mohammadi, Sousan Shalavi, Luciano Giardino, Flavio Palazzi
ABSTRACT
The purpose of the study presented here was to evaluate the effect of Tetraclean, Hypoclean, Chlor-XTRA, 2% chlorhexidine and 6% sodium hypochlorite/17% EDTA as a final irrigant on bacterial leakage of the root canal. One hundred and fifty-five extracted human maxillary central incisors were randomly divided into five experimental groups of 25 teeth each and two control groups of 15 teeth each. The root canals in each group were irrigated with 2 ml of the relevant irrigant between each filing. The root canals in group 5 were irrigated with 5 ml of 17% EDTA at the end of root canal preparation. The teeth in each group were obturated with gutta-percha and AH-26 sealer. Positive control teeth were obturated with a single gutta-percha cone without sealer, and negative controls were obturated in the same way as experimental groups. The coronal portion of each root was placed in contact with inoculum of Enterococcus faecalis in Brain Heart Infusion (BHI) culture media. Findings showed that the mean number of days for bacterial penetration in the Tetraclean group was greater than for other experimental groups. On the other hand, the Chlor-XTRA Vista group showed the fewest mean number of days for bacterial leakage.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, nanopowder, <150 nm particle size, 99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Silver, shot, 1-3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)