Merck
  • Three uncommon KEL alleles in one family with unusual Kell phenotypes explain a 35-year old conundrum.
Three uncommon KEL alleles in one family with unusual Kell phenotypes explain a 35-year old conundrum.
Vox sanguinis (2014-05-06)
V Karamatic Crew, J Poole, N Burton, G Daniels
ABSTRACT
Kell is a complex blood group system comprising 35 antigens. Kell antigens are absent from rare red cells of the Ko (null) phenotype and expressed only weakly in the Kmod phenotype. Molecular analysis of three uncommon KEL alleles elucidated the obscure pattern of inheritance of Kell antigens in one family. Standard serological methods were employed. All exons, flanking intronic sequence and introns 15 and 16 of KEL were sequenced from genomic DNA. cDNA was obtained from erythroid cells cultured from progenitor cells isolated from peripheral blood. The Kmod propositus was heterozygous for two KEL mutations: c.2107G>A, p.Gly703Arg and a synonymous mutation, c.1719C>T, in the codon for p.573Gly. Sequencing of cDNA revealed that c.1719C>T caused skipping of exon 16, resulting in a silent allele. Her KEL:3,-4 brother was heterozygous for KEL*03/04 and c.1719C/T. A synonymous mutation caused complete exon skipping, despite being located 16 bases downstream of the 3′ splice site, resulting in a null KEL allele. The combined effects of two mod alleles, one responsible for KEL3 expression and the other for p.Gly703Arg, were probably responsible for an unexpected KEL:3,-4 phenotype.
Product Description
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Arginine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine 1 M solution
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, 99%, FCC
Supelco
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
SAFC
Glycine
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycine, tested according to Ph. Eur.