Merck
Sulfur-mediated electron shuttling during bacterial iron reduction.
Science (New York, N.Y.) (2014-05-03)
Theodore M Flynn, Edward J O'Loughlin, Bhoopesh Mishra, Thomas J DiChristina, Kenneth M Kemner
ABSTRACT
Microbial reduction of ferric iron [Fe(III)] is an important biogeochemical process in anoxic aquifers. Depending on groundwater pH, dissimilatory metal-reducing bacteria can also respire alternative electron acceptors to survive, including elemental sulfur (S(0)). To understand the interplay of Fe/S cycling under alkaline conditions, we combined thermodynamic geochemical modeling with bioreactor experiments using Shewanella oneidensis MR-1. Under these conditions, S. oneidensis can enzymatically reduce S(0) but not goethite (α-FeOOH). The HS(-) produced subsequently reduces goethite abiotically. Because of the prevalence of alkaline conditions in many aquifers, Fe(III) reduction may thus proceed via S(0)-mediated electron-shuttling pathways.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 99.95%
Iron, foil, 50x50mm, thickness 2mm, as rolled, 99.99+%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 25mm, as drawn, 98+%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 300x300mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%
Iron, foil, 300x300mm, thickness 0.15mm, hard, 99.5%
Iron, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Iron, wire reel, 1000m, diameter 0.125mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.005mm, 99.85%
Iron, tube, 500mm, outside diameter 3.18mm, inside diameter 1.96mm, wall thickness 0.61mm, as drawn, 99.5%
Iron, tube, 200mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%
Iron, wire reel, 20m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1000m, diameter 0.25mm, hard, 99.5%
Iron, wire reel, 250m, diameter 0.25mm, hard, 99.5%