Merck
  • Optimization of oleylamine-Fe3O4/MWCNTs nanocomposite modified GC electrode for electrochemical determination of ofloxacin.
Optimization of oleylamine-Fe3O4/MWCNTs nanocomposite modified GC electrode for electrochemical determination of ofloxacin.
Journal of nanoscience and nanotechnology (2014-04-25)
Deivasigamani Ranjith Kumar, Devaraj Manoj, Jayadevan Santhanalakshmi
ABSTRACT
Iron oxide (Fe3O4) nanoparticles are prepared by nucleation method using oleylamine (OLA) as the capping agent. The OLA capped Fe3O4 nanoparticles (OLA-Fe3O4) are characterized by Fourier Transform Infrared (FT-IR) spectroscopy, X-ray Diffraction (XRD) and High Resolution Transmission Electron Microscopic (HRTEM) analysis. The morphology of the OLA-Fe3O4 nanoparticles is found to be spherical with an average size of 7.5 +/- 0.5 nm. OLA-Fe3O4 nanoparticles are incorporated into the carboxyl functionalized Multi-walled carbon nanotubes (MWCNTs) at different 10, 20, 30, 40 and 50 wt% by ultrasonication. The optimum loading of OLA-Fe3O4 nanoparticles on the MWCNTs was characterized by FTIR, Raman spectroscopy, Field Emission Scanning Electron Microscopy (FESEM) and Linear Sweep Voltammetry (LSV). Finally, the 40wt% of OLA-Fe3O4 nanoparticles on MWCNTs was chosen as the optimum loading and this composition was used for the electrochemical oxidation of ofloxacin (OFX). For determination of OFX in OLA-Fe3O4/MWCNTs modified glassy carbon (GC) electrode show high sensitivity and fast response. The sensitivity of the modified electrode was calculated to be 0.578 microAmicroM(-1) with a detection limit of 0.060 microM. The high sensitivity, wider linear range, good reproducibility and the minimal surface fouling make this OLA-Fe3O4/MWCNTs/GC modified electrode acts as a promising platform for electrochemical determination of OFX.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Iron(III) oxide, nanopowder, <50 nm particle size (BET)
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Iron(III) oxide, ≥99.995% trace metals basis
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
Ofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Supelco
Activated charcoal, powder
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Carbon, foil, 50x50mm, thickness 0.2mm, flexible graphite, 99.8%
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 10mm disks, thickness 0.2mm, flexible graphite, 99.8%