Merck
  • Green and economical synthesis of carbon-coated MoO2 nanocrystallines with highly reversible lithium storage capacity.
Green and economical synthesis of carbon-coated MoO2 nanocrystallines with highly reversible lithium storage capacity.
Journal of nanoscience and nanotechnology (2014-04-18)
Xiaohong Sun, Yifeng Shi, Xiangpeng Fang, Huiming Ji, Xiaolei Li, Shu Cai, Chunming Zheng, Yongsheng Hu
ABSTRACT
Carbon-coated MoO2 nanocrystallines with uniform particle size and carbon-coating morphology have been fabricated by a green and economical hydrothermal route and carbonization process. Glucose here acts as a multifunctional agent, not only as the reducing species to prepare MoO2, but also as the carbonaceous precursor and coating agent to form the carbon-coated and nanoscale MoO2 crystallines. The electrochemical tests demonstrate that the as-synthesized carbon-coated MoO2 nanocrystallines exhibit high capacity and excellent capacity retention as an anode material for lithium-ion batteries. The specific discharge capacity is as high as 790 mA h g(-1) in the first cycle and 730 mA h g(-1) over 50 cycles. The significant enhancement in the electrochemical Li storage performance is attributed to the synergistic effect of the nanocrystallines structure with small particle size and uniform carbon-coating shell, which reduces the diffusion distance for Li-ion and electron, provides high electric conductivity and relieves the volume effect during the cycling.
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Molybdenum, rod, 1000mm, diameter 2.0mm, centerless ground, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.005mm, 99.9%
Molybdenum, wire reel, 100m, diameter 0.2mm, as drawn, 99.95%
Molybdenum, foil, light tested, 25x25mm, thickness 0.004mm, 99.9%
Molybdenum, microfoil, disks, 25mm, thinness 1.0μm, specific density 1022μg/cm2, permanent mylar 3.5μm support, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Molybdenum, foil, light tested, 50x50mm, thickness 0.006mm, 99.9%
Molybdenum, wire reel, 1m, diameter 0.013mm, hard, 99.8+%
Molybdenum, foil, 100x100mm, thickness 0.75mm, annealed, 99.9%
Molybdenum, wire reel, 10m, diameter 0.01mm, hard, 99.8+%
Molybdenum, wire reel, 100m, diameter 0.025mm, hard, 99.95%
Molybdenum, wire reel, 1m, diameter 0.5mm, annealed, 99.95%
Molybdenum, wire reel, 100m, diameter 0.013mm, hard, 99.8+%
Molybdenum, wire reel, 10m, diameter 0.30mm, annealed, 99.95%