Merck
  • Effects of synthesizing parameters on surface roughness and contact angles of ZnO nanowire films.
Effects of synthesizing parameters on surface roughness and contact angles of ZnO nanowire films.
Journal of nanoscience and nanotechnology (2014-04-18)
Weixuan Jing, Bing Wang, Lingling Niu, Zhuangde Jiang, Han Qi, Lujia Chen, Fan Zhou
ABSTRACT
Effects of the synthesizing parameters on the surface roughness and the contact angles of ZnO nanowire films were studied in this paper. ZnO nanowire films were synthesized with the hydrothermal method on glass substrates, and the synthesizing parameters include the concentrations of the growth solution and the seed layer solution, the growth time span as well as the temperature. Atomic force microscopy and scanning electron microscopy were employed respectively to characterize the surface and the profile roughness of ZnO nanowire films. The measurement results by atomic force microscopy were in agreement with that by scanning electron microscopy, hence the former was used for the investigation of aforementioned effects. Relationships between the synthesizing parameters, the surface roughness and the contact angles of ZnO nanowire films were established, revealing that the synthesizing parameters affected significantly not only the surface roughness but also the contact angles of ZnO nanowire films. The results can be used for batch fabrication of ZnO nanowire-based structures and these structures-based sensors in a wide variety of applications.
Product Description
Supelco
Zinc oxide, analytical standard
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, nanowires, Length 1 um
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%