Merck
  • The FXR agonist 6ECDCA reduces hepatic steatosis and oxidative stress induced by ethanol and low-protein diet in mice.
The FXR agonist 6ECDCA reduces hepatic steatosis and oxidative stress induced by ethanol and low-protein diet in mice.
Chemico-biological interactions (2014-04-10)
F A R Lívero, Aline Maria Stolf, Arturo Alejandro Dreifuss, Amanda Leite Bastos-Pereira, Raphaella Chicorski, Liana Gomes de Oliveira, Carlos Eduardo Alves de Souza, Isabella Aviles Fabossi, I S Rabitto, Luiza Helena Gremski, Raílson Henneberg, José Ederaldo Queiroz Telles, Ronald P J Oude Elferink, Alexandra Acco
ABSTRACT
Excessive ethanol consumption can lead to development of hepatic steatosis. Since the FXR receptor regulates adipose cell function and liver lipid metabolism, the aim of this work was to examine the effects of the FXR agonist 6ECDCA on alcoholic liver steatosis development and on oxidative stress induced by ethanol consumption. Swiss mice (n=24) received a low-protein diet (6%) and a liquid diet containing 10% ethanol or water for 6weeks. In the last 15days mice received oral treatment with 6ECDCA (3mgkg(-1)) or 1% tween (vehicle). The experimental groups (n=6) were: water+tween, water+6ECDCA, ethanol+tween and ethanol+6ECDCA. Moreover, as a diet control, we used a basal group (n=6), fed by a normal-proteic diet (23%) and water. After the treatment period, the animals were anesthetized for sample collection to perform plasma biochemistry assays, hepatic oxidative stress assays, hepatic cholesterol and triglycerides measurements, liver histology and hepatic gene expression. Ethanol associated with low-protein diet induced hepatic oxidative stress, increased plasma transaminases and induced hepatic lipid accumulation. Many of these parameters were reversed by the administration of 6ECDCA, including amelioration of lipid accumulation and lipoperoxidation, and reduction of reactive oxygen species. These effects were possibly mediated by regulation of Srebpf1 and FAS gene expression, both reduced by the FXR agonist. Our data demonstrated that 6ECDCA reverses the accumulation of lipids in the liver and decreases the oxidative stress induced by ethanol and low-protein diet. This FXR agonist is promising as a potential therapy for alcoholic liver steatosis.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Chenodeoxycholic acid
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Glutathione S-Transferase from equine liver, lyophilized powder, ≥25 units/mg protein
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®