Merck
  • Pyruvate carboxylase as a sensitive protein biomarker for exogenous steroid chemicals.
Pyruvate carboxylase as a sensitive protein biomarker for exogenous steroid chemicals.
Environmental pollution (Barking, Essex : 1987) (2014-04-01)
Xuefang Liang, Christopher J Martyniuk, Gang Cheng, Jinmiao Zha, Zijian Wang
ABSTRACT
Assessing protein responses to endocrine disrupting chemicals is critical for understanding the mechanisms of chemical action and for the assessment of hazards. In this study, the response of the liver proteome of male rare minnows (Gobiocypris rarus) treated with 17β-estradiol (E2) and females treated with 17α-methyltestosterone (MT) were analyzed. A total of 23 and 24 proteins were identified with differential expression in response to E2 and MT, respectively. Pyruvate carboxylase (PC) was the only common differentially expressed protein in both males and females after E2- and MT-treatments. The mRNA as well as the protein levels of PC were significantly down-regulated compared with that of the controls (p < 0.05). Our results suggest that endocrine disruptors interfere with genes and proteins of the TCA cycle and PC may be a sensitive biomarker of exposure to exogenous steroid chemicals in the liver of fish.
Product Description
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
Pyruvate Carboxylase from bovine liver, buffered aqueous glycerol solution, 5-25 units/mg protein (BCA)
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®