Merck
  • Rilpivirine versus efavirenz with emtricitabine/tenofovir disoproxil fumarate in treatment-naïve HIV-1-infected patients with HIV-1 RNA ≤100,000 copies/mL: week 96 pooled ECHO/THRIVE subanalysis.
Rilpivirine versus efavirenz with emtricitabine/tenofovir disoproxil fumarate in treatment-naïve HIV-1-infected patients with HIV-1 RNA ≤100,000 copies/mL: week 96 pooled ECHO/THRIVE subanalysis.
AIDS patient care and STDs (2014-03-26)
Georg Behrens, Bart Rijnders, Mark Nelson, Chloe Orkin, Calvin Cohen, Anthony Mills, Richard A Elion, Simon Vanveggel, Marita Stevens, Laurence Rimsky, David Thorpe, Matthew Bosse, Kirsten White, Lijie Zhong, Jennifer DeMorin, Susan K Chuck
ABSTRACT
The once daily, single-tablet regimen (STR) combining rilpivirine (RPV), emtricitabine (FTC), and tenofovir disoproxil fumarate (TDF) provides a simplified treatment option for antiretroviral therapy (ART)-naïve patients with baseline HIV-1 RNA (BLVL) of ≤100,000 copies/mL. The aim of this analysis is to compare long-term efficacy, safety, and tolerability of RPV+FTC/TDF vs. efavirenz (EFV)+FTC/TDF as individual components in subjects with BLVL ≤100,000 copies/mL. Week 96 efficacy and safety data from subjects with BLVL ≤100,000 copies/mL, who received daily RPV 25 mg or EFV 600 mg with FTC/TDF in the phase 3, randomized, double-blind, double-dummy, active-controlled, registrational trials ECHO and THRIVE, were analyzed. Virologic response was evaluated by intent-to-treat, time to loss of virological response (ITT-TLOVR), and Snapshot algorithms. Through Week 96, RPV+FTC/TDF demonstrated non-inferior efficacy to EFV+FTC/TDF (84% vs. 81%, respectively; ITT-TLOVR) in 543 subjects with BLVL ≤100,000 copies/mL, and overall rates of virologic failure (VF) were 5.9% vs. 2.4%, respectively. Resistance development was lower in Year 2 than Year 1. Subjects in both arms with suboptimal adherence (≤95%) had lower virologic responses (63% vs. 62%, respectively). Treatment with RPV+FTC/TDF was associated with significantly fewer treatment-related adverse events (AEs), grade 2-4 AEs, neurological and psychiatric AEs (including dizziness and abnormal dreams/nightmares), and rash. Additionally, grade 2-4 treatment-emergent laboratory abnormalities and grade 1-3 lipid abnormalities were significantly less common with RPV+FTC/TDF than EFV+FTC/TDF. RPV+FTC/TDF demonstrated non-inferior efficacy to EFV+FTC/TDF in ART-naïve subjects with BLVL ≤100,000 copies/mL and was associated with a higher rate of VF but a more favorable safety and tolerability profile through Week 96.
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Adenine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Supelco
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material