Merck
  • Aromatase transgenic upregulation modulates basal cardiac performance and the response to ischemic stress in male mice.
Aromatase transgenic upregulation modulates basal cardiac performance and the response to ischemic stress in male mice.
American journal of physiology. Heart and circulatory physiology (2014-03-13)
James R Bell, Gabriel B Bernasochi, Upasna Varma, Wah Chin Boon, Stuart J Ellem, Gail P Risbridger, Lea M D Delbridge
ABSTRACT
Estrogen in females is conventionally considered a cardioprotective influence, but a role for estrogen in male cardioprotection has yet to be defined. Estrogen biosynthesis from testosterone is regulated by aromatase. Aromatase has recently been shown to be expressed in the adult heart, although little is known about its involvement in the regulation of myocardial function and stress responses. The goal of this study was to determine whether upregulation of tissue aromatase expression could improve ischemic resilience in male hearts. Isolated hearts from male transgenic aromatase-overexpressing (AROM(+); high estrogen, low testosterone) mice and wild-type (WT) mice (12 wk) were Langendorff perfused and subjected to ischemia-reperfusion (25 min ischemia and 60 min of reperfusion). Basal systolic function was lower in AROM(+) hearts (dP/dtmax: 4,121 ± 255 vs. 4,992 ± 283 mmHg/s, P < 0.05) and associated with augmented Akt phosphorylation, consistent with a suppressor action of estrogen on contractility. Ischemic contracture was attenuated in AROM(+) hearts (43 ± 3 vs. 55 ± 4 mmHg, P < 0.05), yet AROM(+) hearts were more arrhythmic in early reperfusion. At the end of 60 min of reperfusion, AROM(+) systolic functional recovery was lower (left ventricular developed pressure: 39 ± 6 vs. 56 ± 5 %basal, P < 0.05) and diastolic dysfunction was accentuated (36 ± 4 vs. 24 ± 2 mmHg, P < 0.05). This is the first study to show that in vivo aromatase upregulation modulates basal cardiac performance and the response to ischemic stress. These data suggest that while chronic exposure to enhanced estrogenic influence may have benefits in limiting ischemic contracture severity, acute functional recovery in reperfusion is compromised. A temporally targeted, tissue-specific intervention combining aromatase treatment with inotropic support may offer therapeutic potential for men and women.
Product Description
Testosterone, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Supelco
Testosterone, VETRANAL®, analytical standard
Pricing and availability is not currently available.