Merck
  • Radiotherapy for patients with isolated local recurrence of primary resected pancreatic cancer. Prolonged disease-free interval associated with favorable prognosis.
Radiotherapy for patients with isolated local recurrence of primary resected pancreatic cancer. Prolonged disease-free interval associated with favorable prognosis.
Strahlentherapie und Onkologie : Organ der Deutschen Rontgengesellschaft ... [et al] (2014-03-07)
Akira Nakamura, Satoshi Itasaka, Kyoichi Takaori, Yoshiya Kawaguchi, Keiko Shibuya, Michio Yoshimura, Yukinori Matsuo, Takashi Mizowaki, Shinji Uemoto, Masahiro Hiraoka
ABSTRACT
To evaluate the treatment outcomes of radiotherapy and prognostic factors for recurrent pancreatic cancer. The study comprised 30 patients who developed a locoregional recurrence of primarily resected pancreatic cancer and received radiotherapy between 2000 and 2013 with a median dose of 54 Gy (range, 39-60 Gy). Concurrent chemotherapy included gemcitabine for 18 patients and S-1 for seven patients. The treatment outcomes and prognostic factors were retrospectively analyzed. The median follow-up after radiotherapy was 14.6 months. The 1-year overall survival, local control, and progression-free survival rates were 69%, 67%, and 32%, respectively. The median overall survival and progression-free survival rates were 15.9 and 6.9 months, respectively. Tumor marker reduction and ≥ 50% reduction were observed in 18 and two patients, respectively. Of the seven patients who exhibited pain symptoms, four and two patients were partly and completely relieved, respectively. Late grade 3 ileus and gastroduodenal bleeding were observed in one patient each. Among the clinicopathological factors evaluated, only a disease-free interval of greater than 18.9 months exhibited a significant association with improved overall survival (p = 0.017). Radiotherapy for isolated locally recurrent pancreatic cancer resulted in encouraging local control, overall survival, and palliative effects with mild toxicity, particularly in patients with a prolonged disease-free interval. This treatment strategy should be prospectively evaluated.
Sigma-Aldrich
2′-Deoxycytidine, ≥99% (HPLC)
Sigma-Aldrich
Tegafur, ≥98% (HPLC), powder