Merck
  • Accumulation of heavy metals in different body tissues of Atlantic salmon, Salmo salar L., exposed to a model mixture (Cu, Zn, Ni, Cr, Pb, Cd) and singly to nickel, chromium, and lead.
Accumulation of heavy metals in different body tissues of Atlantic salmon, Salmo salar L., exposed to a model mixture (Cu, Zn, Ni, Cr, Pb, Cd) and singly to nickel, chromium, and lead.
Bulletin of environmental contamination and toxicology (2014-03-04)
Gintaras Svecevičius, Gintarė Sauliutė, Raimondas Leopoldas Idzelis, Joana Grigelevičiūtė
ABSTRACT
One-year-old Atlantic salmon smolts were exposed for 2 weeks either to six priority heavy metal mixture or to Ni, Pb, and Cr singly at a concentration corresponding to Lithuanian inland water standards: Cu - 0.01, Zn - 0.1, Ni - 0.01, Cr - 0.01, Cd - 0.005 and Pb - 0.005 mg/L, respectively under semi-static conditions. The presence of metal mixture in the water only partly (by 50 %) affected the accumulation of single metals in body tissues (muscle, gills, liver and kidneys) probably due to the synergistic interactions among metals. Although metal concentrations increased in most cases, only Pb exceeded recommended level for human consumption (0.2 Pb mg/L) by 1.1-fold to 2.1-fold.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Nickel, tube, 500mm, outside diameter 8.4mm, inside diameter 8.15mm, wall thickness 0.125mm, hard, 99.5%
Nickel, foil, 10mm disks, thickness 0.003mm, 99.95%
Nickel, foil, 25x25mm, thickness 0.125mm, annealed, 99.98%
Nickel, foil, 0.5m coil, thickness 0.004mm, hard, 99.98%
Nickel, foil, 4mm disks, thickness 0.007mm, 99.95%
Nickel, foil, 4mm disks, thickness 0.025mm, as rolled, 100%
Nickel, foil, 2m coil, thickness 0.125mm, coil width 50mm, annealed, 99%
Nickel, foil, 10m coil, thickness 0.125mm, coil width 50mm, annealed, 99%
Lead, foil, 2m coil, thickness 0.12mm, coil width 25.4mm, as rolled, 99.95%
Nickel, foil, 2m coil, thickness 0.20mm, coil width 1mm, annealed, 99%
Nickel, foil, 25x25mm, thickness 1.0mm, annealed, 99.98%
Nickel, foil, 25x25mm, thickness 0.125mm, annealed, 99%
Nickel, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.15mm, coil width 12mm, annealed, 99%
Nickel, wire reel, 10m, diameter 0.01mm, as drawn, 99.98%
Nickel, foil, 0.5m coil, thickness 0.125mm, annealed, 99%
Nickel, foil, 4mm disks, thickness 0.003mm, 99.95%