Merck
  • Ethanol disrupts intestinal epithelial tight junction integrity through intracellular calcium-mediated Rho/ROCK activation.
Ethanol disrupts intestinal epithelial tight junction integrity through intracellular calcium-mediated Rho/ROCK activation.
American journal of physiology. Gastrointestinal and liver physiology (2014-02-22)
Elhaseen Elamin, Ad Masclee, Jan Dekker, Daisy Jonkers
ABSTRACT
Evidence indicates that ethanol-induced intestinal barrier dysfunction and subsequent endotoxemia plays a key role in the pathogenesis of alcoholic liver disease. Recently, it has been demonstrated that ethanol induces RhoA kinase activation in intestinal epithelium, thereby disrupting barrier integrity. In this study, the role of a rise in intracellular calcium concentration ([Ca(2+)]i) in ethanol-induced Rho-associated coiled coil-forming kinase (Rho/ROCK) activation and barrier disruption was investigated in Caco-2 cell monolayers. Treatment of Caco-2 monolayers with 40 mmol/l ethanol induced [Ca(2+)]i release as indicated by increased relative fluorescent units of Fluo-3 from 0.06 ± 0.02 to 2.27 ± 1.96 (P < 0.0001). Pretreatment with 1,2-bis(2-aminophenoxy) ethane-N,N,N',N'-tetraacetic acid (BAPTA-AM) completely inhibited the release, whereas the inositol 1,4,5-triphosphate receptor (IP3R)-antagonist, Xestospongin C, partially inhibited the ethanol-induced [Ca(2+)]i release (from 2.27 ± 1.96 to 0.03 ± 0.01; P < 0.0001 and from 2.27 ± 1.96 to 1.19 ± 1.80; P < 0.001, respectively). The rise in [Ca(2+)]i was paralleled with increased intestinal permeability, which could be attenuated by either BAPTA-AM or Xestospongin C. Furthermore, ethanol induced Rho/ROCK activation, as indicated by increased phosphorylation of myosin-binding subunit, which could be prevented either by BAPTA, Xestospongin C, or the specific Rho/ROCK inhibitor Y27632. Finally, inhibition of Rho/ROCK kinase by Y27632 ameliorated the ethanol-induced redistribution of zonula occluden-1, adherens junction proteins including E-cadherin and β-catenin, and also disorganization of F-actin. These findings suggest that ethanol-induced [Ca(2+)]i release, mediated by stimulating IP3R-gated Ca(2+) channel, activates Rho/ROCK in Caco-2 cells, thereby contributing to ethanol-induced intestinal barrier dysfunction.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, for residue analysis
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology