Merck
  • In a nongenomic action, steroid hormone 20-hydroxyecdysone induces phosphorylation of cyclin-dependent kinase 10 to promote gene transcription.
In a nongenomic action, steroid hormone 20-hydroxyecdysone induces phosphorylation of cyclin-dependent kinase 10 to promote gene transcription.
Endocrinology (2014-02-13)
Wen Liu, Mei-Juan Cai, Jin-Xing Wang, Xiao-Fan Zhao
ABSTRACT
The insect steroid hormone 20-hydroxyecdysone (20E) regulates gene transcription via a genomic pathway by forming a transcription complex that binds to DNA with the help of the chaperone proteins, heat shock proteins (Hsps) Hsc70 and Hsp90. However, the nongenomic mechanisms by which 20E regulates gene expression remain unclear. In this study, we found that 20E regulated the phosphorylation of serine/threonine protein kinase cyclin-dependent kinase 10 (CDK10) through a nongenomic pathway to mediate gene transcription in the lepidopteran Helicoverpa armigera. The down-regulation of CDK10 by RNA interference in larvae and the epidermal cell line delayed development and suppressed 20E-induced gene transcription. CDK10 was localized to the nucleus via its KKRR motif, and this nuclear localization and the ATPase motif were necessary for the efficient expression of the 20E-inducible gene. The rapid phosphorylation of CDK10 was induced by 20E, whereas it was repressed by the inhibitors of G-protein-coupled receptors, phospholipase C, and Ca²⁺ channels. Phosphorylated CDK10 exhibited increased interactions with Hsps Hsc70 and Hsp90 and then promoted the interactions between Hsps and ecdysone receptor EcRB1 and the binding of the Hsps-EcRB1 complex to the 20E response element for the regulation of gene transcription. CDK10 depletion suppressed the formation of the Hsps-EcRB1 complex at the hormone receptor 3 promoter. These results suggest that 20E induces CDK10 phosphorylation via a nongenomic pathway to regulate gene transcription in the nucleus.
Sigma-Aldrich
Suramin sodium salt, ≥98% (TLC)
Sigma-Aldrich
20-Hydroxyecdysone, ≥93% (HPLC), powder
Sigma-Aldrich
Pyrazole, 98%
Sigma-Aldrich
Flunarizine dihydrochloride, ≥98% (TLC)
Flunarizine dihydrochloride, European Pharmacopoeia (EP) Reference Standard