Merck
  • Resistant starch and arabinoxylan augment SCFA absorption, but affect postprandial glucose and insulin responses differently.
Resistant starch and arabinoxylan augment SCFA absorption, but affect postprandial glucose and insulin responses differently.
The British journal of nutrition (2014-02-11)
Anne Krog Ingerslev, Peter Kappel Theil, Mette Skou Hedemann, Helle Nygaard Lærke, Knud Erik Bach Knudsen
ABSTRACT
The effects of increased colonic fermentation of dietary fibres (DF) on the net portal flux (NPF) of carbohydrate-derived metabolites (glucose, SCFA and, especially, butyrate), hormones (insulin, C-peptide, glucagon-like peptide 1 and glucose-dependent insulinotropic peptide) and NEFA were studied in a healthy catheterised pig model. A total of six pigs weighing 59 (SEM 1·6) kg were fitted with catheters in the mesenteric artery and in the portal and hepatic veins, and a flow probe around the portal vein, and included in a double 3 × 3 cross-over design with three daily feedings (at 09.00, 14.00 and 19.00 hours). Fasting and 5 h postprandial blood samples were collected after 7 d adaptation to each diet. The pigs were fed a low-DF Western-style control diet (WSD) and two high-DF diets (an arabinoxylan-enriched diet (AXD) and a resistant starch-enriched diet (RSD)). The NPF of insulin was lower (P= 0·04) in AXD-fed pigs (4·6 nmol/h) than in RSD-fed pigs (10·5 nmol/h), despite the lowest NPF of glucose being observed in RSD-fed pigs (203 mmol/h, P= 0·02). The NPF of total SCFA, acetate, propionate and butyrate were high, intermediate and low (P< 0·01) in AXD-, RSD- and WSD-fed pigs, respectively, with the largest relative increase being observed for butyrate in response to arabinoxylan supplementation. In conclusion, the RSD and AXD had different effects on the NPF of insulin and glucose, suggesting different impacts of arabinoxylan and resistant starch on human health.
Sigma-Aldrich
Starch from corn, practical grade
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from potato, Soluble