Merck
Inhibitory effects of PLAP-1/asporin on periodontal ligament cells.
Journal of dental research (2014-01-24)
T Kajikawa, S Yamada, T Tauchi, T Awata, S Yamaba, C Fujihara, S Murakami
ABSTRACT
PLAP-1/asporin is an extracellular matrix protein that is predominantly expressed in the human periodontal ligament (PDL) and has an aspartic acid (D) repeat polymorphism in its N-terminal region. In this study, we hypothesized that the D repeat polymorphism of PLAP-1/asporin may affect the physiological functions of periodontal ligaments. We established periodontal ligament cell lines transfected with the D13- or D14-PLAP-1 gene. Alkaline phosphatase staining and alizarin red staining revealed that the cytodifferentiation of the D14-PLAP-1-expressing PDL cells was more repressed compared with that of the D13-PLAP-1-expressing cells. Furthermore, the D14-PLAP-1-expressing cells inhibited BMP-2-induced cytodifferentiation more strongly than did the D13-PLAP-1-expressing cells. Western blotting analysis and luciferase assay revealed that D14-PLAP-1 suppressed BMP-2 signal transduction more efficiently than did D13-PLAP-1, and co-immunoprecipitation demonstrated the stronger affinity of the D14-PLAP-1 protein to BMP-2 compared with the D13-PLAP-1 protein. Analysis of these data suggests that the D repeat polymorphism of PLAP-1/asporin has a significant influence on the functions of PDL cells.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Alizarin, Dye content 97 %
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
SAFC
L-Aspartic acid
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)