Merck
  • Tyrosine kinase inhibitor treatments in patients with metastatic thyroid carcinomas: a retrospective study of the TUTHYREF network.
Tyrosine kinase inhibitor treatments in patients with metastatic thyroid carcinomas: a retrospective study of the TUTHYREF network.
European journal of endocrinology (2014-01-16)
Marie-Hélène Massicotte, Maryse Brassard, Médéric Claude-Desroches, Isabelle Borget, Françoise Bonichon, Anne-Laure Giraudet, Christine Do Cao, Cécile N Chougnet, Sophie Leboulleux, Eric Baudin, Martin Schlumberger, Christelle de la Fouchardière
ABSTRACT
Tyrosine kinase inhibitors (TKIs) are used to treat patients with advanced thyroid cancers. We retrospectively investigated the efficacy of TKIs administered outside of clinical trials in metastatic sites or locally advanced thyroid cancer patients from five French oncology centers. THERE WERE 62 PATIENTS (37 MEN, MEAN AGE: 61 years) treated with sorafenib (62%), sunitinib (22%), and vandetanib (16%) outside of clinical trials; 22 had papillary, five had follicular, five had Hürthle cell, 13 had poorly differentiated, and 17 had medullary thyroid carcinoma (MTC). Thirty-three, 25, and four patients were treated with one, two, and three lines of TKIs respectively. Primary endpoints were objective tumor response rate and progression-free survival (PFS). Sequential treatments and tumor response according to metastatic sites were secondary endpoints. Among the 39 sorafenib and 12 sunitinib treatments in differentiated thyroid carcinoma (DTC) patients, partial response (PR) rate was 15 and 8% respectively. In the 11 MTC patients treated with vandetanib, 36% had PR. Median PFS was similar in second-line compared with first-line sorafenib or sunitinib therapy (6.7 vs 7.0 months) in DTC patients, but there was no PR with second- and third-line treatments. Bone and pleural lesions were the most refractory sites to treatment. This is the largest retrospective study evaluating TKI therapies outside of clinical trials. DTC patients treated with second-line therapy had stable disease as best response, but had a similar median PFS compared with the first-line treatment.
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard