- Cesium accumulation of Rhodococcus erythropolis CS98 strain immobilized in hydrogel matrices.
Cesium accumulation of Rhodococcus erythropolis CS98 strain immobilized in hydrogel matrices.
Journal of bioscience and bioengineering (2013-11-05)
Takayuki Takei, Mika Yamasaki, Masahiro Yoshida
PMID24183457
ABSTRACT
Agarose gels were superior to calcium-alginate gels for immobilizing Rhodococcus erythropolis CS98 strain to remove cesium from water. Suitable incubation time of the immobilized cells in cesium solutions, cell number in the gels and volume ratio of the cesium solution to the gels for efficient cesium removal were identified.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Sepharose™ 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor