Merck
  • Effects of Mn-DPDP and manganese chloride on hemodynamics and glucose tolerance in anesthetized rats.
Effects of Mn-DPDP and manganese chloride on hemodynamics and glucose tolerance in anesthetized rats.
Acta radiologica (Stockholm, Sweden : 1987) (2013-08-22)
Xiang Gao, Monica Sandberg, My Quach, Birgitta Bodin, Lars Johansson, Leif Jansson
ABSTRACT
Previous studies have demonstrated that magnetic resonance imaging may be a method of choice to visualize transplanted pancreatic islets. However, contrast agents may interfere with microcirculation and affect graft function. To evaluate the effects manganese-containing contrast media on regional blood flow and glucose tolerance. Anesthetized rats were injected intravenously with MnCl2 (10 µM/kg body weight) or Mn-DPDP (Teslascan™; 5 µM/kg body weight). Blood flow measurements were made with a microsphere technique 10 min later. In separate animals vascular arteriolar reactivity in isolated, perfused islets was examined. Furthermore, an intraperitoneal glucose tolerance test was performed in separate rats. Glucose tolerance was unaffected by both agents. No changes in regional blood flow were seen after administration of Mn-DPDP, except for an increase in arterial liver blood flow. MnCl2 increased all blood flow values except that of the kidney. MnCl2, but not Mn-DPDP, caused a vasoconstriction in isolated rat islet arterioles but only at very high doses. Mn-DPDP administration does not affect glucose tolerance or regional blood flow, besides an increase in arterial hepatic blood flow, and may therefore be suitable for visualization of islets.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, 98.5-101.5%, BioUltra
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
USP
Edetate disodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Disodium edetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, for molecular biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sodium calcium edetate, European Pharmacopoeia (EP) Reference Standard