Merck
  • Tequila volatile characterization and ethyl ester determination by solid phase microextraction gas chromatography/mass spectrometry analysis.
Tequila volatile characterization and ethyl ester determination by solid phase microextraction gas chromatography/mass spectrometry analysis.
Journal of agricultural and food chemistry (2004-09-18)
Belinda Vallejo-Cordoba, Aarón Fernando González-Córdova, María del Carmen Estrada-Montoya
ABSTRACT
Solid phase microextraction (SPME) and gas chromatography were used for tequila volatile characterization and ethyl ester quantitation. Several factors determined the differences in tequila volatile profiles obtained by the SPME technique, namely, sampling mode, fiber coating, and fiber exposure time. Each of these factors determined the most suitable conditions for the analysis of volatile profiles in tequila. Volatile extraction consisted of placing 40 mL of tequila in a sealed vial kept at 40 degrees C. A poly(dimethylsiloxane) fiber was immersed in the liquid for 60 min and desorbed for 5 min into the gas chromatograph. The identified volatiles by mass spectrometry were mainly alcohols, esters, and ketones. The calibration curves for ethyl hexanoate, octanoate, and decanoate followed linear relationships with highly significant (p < 0.001) determination coefficients (R2 = 0.99). The coefficients of variation of less than 10% for ethyl ester concentrations indicated that the technique was reproducible. The limits of quantitation for ethyl esters were 0.05 parts per million, which were below the concentration range (0.27-15.03 ppm) found for different tequila samples. Quantitative differences in ethyl esters were found for the four most commonly known tequila types: silver, gold, aged, and extra-aged.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler