Merck
  • Possible interest of various sample transfer techniques for fast gas chromatography-mass spectrometric analysis of true onion volatiles.
Possible interest of various sample transfer techniques for fast gas chromatography-mass spectrometric analysis of true onion volatiles.
Journal of chromatography. A (2000-11-28)
I Arnault, N Mondy, F Cadoux, J Auger
ABSTRACT
We improved GC-MS analysis of onion volatiles by comparing organic solvent partition with solid-phase microextraction (SPME) following cryo-trapping isolation and by comparing the same extraction methods on direct onion juice. Cryotrapping produces very small quantities of volatiles and therefore is not a suitable extraction method for GC-MS analysis. We confirm that SPME accelerates the degradation of labile thiosulfinates but the lacrymatory factor remains intact. The identification of Allium thiosulfinates is only obtained on juice extracted by diethyl ether using a fast GC-MS analysis on a 10 m X 0.3 mm column of 4 microm coating, with routine splitless injection. The lacrymatory factor is best analysed directly on fresh onion juice by SPME with the same chromatographic conditions. To characterise and to quantify all the true onion volatiles, we propose to analyse the same sample by successive SPME-GC-MS and solvent extraction-GC-MS.
Product Description
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder