Merck
  • Collagen functionalized bioactive nanofiber matrices for osteogenic differentiation of mesenchymal stem cells: bone tissue engineering.
Collagen functionalized bioactive nanofiber matrices for osteogenic differentiation of mesenchymal stem cells: bone tissue engineering.
Journal of biomedical nanotechnology (2014-04-18)
Yixing Cheng, Daisy Ramos, Paul Lee, Danni Liang, Xiaojun Yu, Sangamesh G Kumbar
ABSTRACT
Scaffold architecture, surface topography, biochemical and mechanical cues have been shown to significantly improve cellular events and in vivo tissue regeneration. Specifically electrospun nanofiber matrices have gained tremendous interest due to their intrinsic structural resemblance to native tissue extracellular matrix (ECM). The present study reports on the electrospun nanofiber matrices of polycaprolactone (PCL)-chitosan (CS) blends and effect of type I collagen surface functionalization in regulating rat bone marrow derived stromal cells (rBMSCs) differentiation into osteogenic lineage. Collagen was covalently attached to blend nanofibers via carbodiimide (EDC) coupling. Bead-free smooth nanofibers (diameter-700-850 nm) obtained at the optimized conditions of polymer concentration and electrospinning parameters were used for the study. EDC collagen coupling resulted in 0.120+/-0.016 micro g of collagen immobilization onto a 1 cm2 area of the PCL/CS nanofibers, which was 2.6-folds higher than the amount of collagen that can be retained by physical adsorption. Significantly improved rBMSCs adhesion, spreading, proliferation and osteogenic differentiation was observed on the collagen functionalized COL-PCULCS nanofiber matrices as compared to control groups. Osteogenic phenotypic markers such as alkaline phosphatase (ALP) activity and mineralization were found to be significantly higher on COL-PCL/CS nanofiber matrices as compared to controls. Elevated gene expression profiles of osteogenic markers such as osteocalcin (0CN), osteopontin (OPN) and ALP further corroborate the osteoinductive nature of the collagen functionalized PCL/CS nanofiber matrices. These fiber matrices and modification techniques could be extended to other scaffold systems for tissue engineering applications.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host