Merck
  • Comparative safety of the antifouling compound butenolide and 4,5-dichloro-2-n-octyl-4-isothiazolin-3-one (DCOIT) to the marine medaka (Oryzias melastigma).
Comparative safety of the antifouling compound butenolide and 4,5-dichloro-2-n-octyl-4-isothiazolin-3-one (DCOIT) to the marine medaka (Oryzias melastigma).
Aquatic toxicology (Amsterdam, Netherlands) (2014-03-04)
Lianguo Chen, Rui Ye, Ying Xu, Zhaoming Gao, Doris W T Au, Pei-Yuan Qian
ABSTRACT
This study evaluated the potential adverse effects of butenolide, a promising antifouling compound, using the marine medaka (Oryzias melastigma), a model fish for marine ecotoxicology. The active ingredient used in the commercial antifoulant SeaNine 211, 4,5-dichloro-2-n-octyl-4-isothiazolin-3-one (DCOIT) was employed as the positive control. Adult marine medaka (4-month-old) were exposed to various concentrations of butenolide or DCOIT for 28 days and then depurated in clean seawater for 14 days (recovery). A suite of sensitive biomarkers, including hepatic oxidative stress, neuronal signal transmission, endocrine disruption, and reproductive function, was used to measure significant biological effects induced by the chemicals. Compared to DCOIT, chronic exposure to butenolide induced a lower extent of oxidative stress in the liver of male and female medaka. Furthermore, butenolide-exposed fish could recover faster from oxidative stress than fish exposed to DCOIT. Regarding neurotransmission, DCOIT significantly inhibited acetylcholinesterase (AChE) activity in the brain of both male and female medaka, whereas this was not significant for butenolide. In addition, plasma estradiol (E2) level was elevated and testosterone (T) level was decreased in male medaka exposed to DCOIT. This greatly imbalanced sex hormones ratio (E2/T) in exposed males, indicating that DCOIT is a potent endocrine disruptive chemical. In contrast, butenolide induced only moderate effects on sex hormone levels in exposed males, which could be gradually recovered during depuration. Moreover, the endocrine disruptive effect induced by butenolide did not affect normal development of offspring. In contrast, DCOIT-exposed fish exhibited a decrease of egg production and impaired reproductive success. Overall, the above findings demonstrated that chronic exposure to butenolide induced transient, reversible biological effect on marine medaka, while DCOIT could impair reproductive success of fish, as evident by clear alterations of the E2/T ratio. The relatively low toxicity of butenolide on marine biota highlights its promising application in the antifouling industry. The present findings also emphasize gender difference in fish susceptibility to chemical treatment (male>female), which is an important consideration for ecological risk assessment.
Product Description
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2(5H)-Furanone, 98%
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
γ-Butyrolactone, analytical standard