Merck
  • Injection of phosphatidylcholine and deoxycholic acid regulates gene expression of lipolysis-related factors, pro-inflammatory cytokines, and hormones on mouse fat tissue.
Injection of phosphatidylcholine and deoxycholic acid regulates gene expression of lipolysis-related factors, pro-inflammatory cytokines, and hormones on mouse fat tissue.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2013-08-06)
Tae Joon Won, Yunsung Nam, Ho Sung Lee, Sujin Chung, Jong Hyuk Lee, Yoon Hee Chung, Eon Sub Park, Kwang Woo Hwang, Ji Hoon Jeong
ABSTRACT
Injection of phosphatidylcholine (PC) and deoxycholic acid (DA) preparation is widely used as an alternative to liposuction for the reduction of subcutaneous fat. Nevertheless, its physiological effects and mechanism of action are not yet fully understood. In this report, PC and deoxycholic acid (DA) were respectively injected into adipose tissue. PC decreased tissue mass on day 7, but DA did not. On the other hand, a decrement of DNA mass was observed only in DA-injected tissue on day 7. Both PC and DA reduced the mRNA expression of adipose tissue hormones, such as adiponectin, leptin, and resistin. In lipolysis-related gene expression profiles, PC increased hormone-sensitive lipase (HSL) transcription and decreased the expression other lipases, perilipin, and the lipogenic marker peroxisome proliferator-activated receptor-γ (PPARγ); DA treatment diminished them all, including HSL. Meanwhile, the gene expression of pro-inflammatory cytokines and a chemokine was greatly elevated in both PC-injected and DA-injected adipose tissue. Microscopic observation showed that PC induced lipolysis with mild PMN infiltration on day 7. However, DA treatment did not induce lipolysis but induced much amount of PMN infiltration. In conclusion, PC alone might induce lipolysis in adipose tissue, whereas DC alone might induce tissue damage.
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Cholesterol Esterase from bovine pancreas, lyophilized powder, ≥200 units/g protein
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas sp., lyophilized powder, ≥200,000 units/g protein
Sigma-Aldrich
Cholesterol Esterase from Pseudomonas fluorescens, lyophilized powder, ≥10,000 units/g protein
Sigma-Aldrich
Cholesterol Esterase from porcine pancreas, lyophilized, powder, white, ~35 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells