Merck
Structural basis for bending of organic crystals.
Chemical communications (Cambridge, England) (2005-08-03)
C Malla Reddy, Ravi C Gundakaram, Srinivas Basavoju, Michael T Kirchner, K Anantha Padmanabhan, Gautam R Desiraju
ABSTRACT
Bending is observed in organic crystals when the packing is anisotropic in such a way that strong and weak interaction patterns occur in nearly perpendicular directions.
Sigma-Aldrich
2-(Methylthio)pyridine-3-carboxylic acid, 98%