Merck
  • Enzyme-assisted synthesis and structural characterization of nitrocatechol glucuronides.
Enzyme-assisted synthesis and structural characterization of nitrocatechol glucuronides.
Bioconjugate chemistry (1999-01-20)
L Luukkanen, I Kilpeläinen, H Kangas, P Ottoila, E Elovaara, J Taskinen
ABSTRACT
Enzyme-assisted synthesis and characterization are described for 3-O-beta-D-glucuronides 1b-4b of the aglycons E- and Z-2-cyano-N, N-diethyl-3-(3,4-dihydroxy-5-nitrophenyl)propenamide (entacapone), 1a and 2a, respectively, 3-(3,4-dihydroxy-5-nitrobenzylidene)-2, 4-pentanedione (nitecapone) 3a and 4'-methyl-3, 4-dihydroxy-5-nitrobenzophenone (tolcapone) 4a, and 1-o- and 2-o-glucuronides 5b and 6b of the aglycon 1, 2-dihydroxy-4-nitrobenzene 5a. Liver microsomes from rats pretreated with Aroclor 1254 were used as catalyst in the synthesis. Glucuronidation was regio- and stereoselective in the case of 1a-4a; only one product was observed by HPLC, HPTLC, and NMR. The glucuronidation of 1,2-dihydroxy-4-nitrobenzene 5a resulted in equal amounts of 1-O-beta-D- and 2-O-beta-D-glucuronides. Purification of the crude products by C18 solid-phase extraction and/or flash chromatography gave compounds 1b-6b in 38-98% yields (50-84 mg). The structures of the glucuronides were characterized on the basis of UV and IR spectra and confirmed with FAB-MS and NMR spectroscopy.
Sigma-Aldrich
Nitecapone, ≥98% (HPLC)