Merck
  • Highly sensitive and selective high-performance liquid chromatography method for bioequivalence study of cefpodoxime proxetil in rabbit plasma via fluorescence labeling of its active metabolite.
Highly sensitive and selective high-performance liquid chromatography method for bioequivalence study of cefpodoxime proxetil in rabbit plasma via fluorescence labeling of its active metabolite.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-07-31)
Sameh Ahmed, Hanaa M Abdel-Wadood, Niveen A Mohamed
ABSTRACT
Cefpodoxime proxetil (CFP), a broad-spectrum third-generation cephalosporin, has been used most widely in the treatment of respiratory and urinary tract infections. For bioequivalence study of CFP in rabbit plasma, it was necessary to develop a highly sensitive and selective high-performance liquid chromatographic (HPLC) method with fluorescence (FL) detection. The pre-column labeling of cefpodoxime acid (CFA) (active metabolite) with an efficient benzofurazan type fluorogenic reagent, 4-N,N-dimethyl aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole (DBD-F) was carried out in the present study in 100mM borate buffer (pH=8.5) at 50°C for 15min. The obtained fluorescent products were separated on C18 column with an isocratic elution of the mobile phase, which consists of 10mM phosphate buffer (pH=3.5)/CH3CN (70:30, v/v). The fluorescent product (DBD-CFA) was detected fluorimetrically at 556nm with an excitation wavelength of 430nm. Cefotaxime sodium was used as internal standard. The method was validated according to the requirements of US-FDA guidelines. The correlation coefficient of 0.999 was obtained in the concentration ranges of 10-1000ngmL(-1). The limits of detection and quantification (S/N=3) were 3 and 10ngmL(-1), respectively. Plasma CFA levels were successfully determined in rabbit with satisfactory precision and accuracy. The proposed HPLC-FL method was successfully applied to study bioequivalence in rabbits for two formulations of different brands contained CFP (prodrug) in a randomized, two-way, single-dose, crossover study and all pharmacokinetic parameters for the two formulations were assessed.
Product Description
Cefpodoxime proxetil for peak identification, European Pharmacopoeia (EP) Reference Standard
Cefpodoxime proxetil for impurity H identification, European Pharmacopoeia (EP) Reference Standard
USP
Cefpodoxime proxetil, United States Pharmacopeia (USP) Reference Standard
Cefpodoxime proxetil, European Pharmacopoeia (EP) Reference Standard