Merck
  • Triphala and its active constituent chebulinic acid are natural inhibitors of vascular endothelial growth factor-a mediated angiogenesis.
Triphala and its active constituent chebulinic acid are natural inhibitors of vascular endothelial growth factor-a mediated angiogenesis.
PloS one (2012-09-01)
Kai Lu, Debanjan Chakroborty, Chandrani Sarkar, Tingting Lu, Zhiliang Xie, Zhongfa Liu, Sujit Basu
ABSTRACT
Triphala churna (THL) is a combination of three fruits that has been used for many years in India for the treatment of various diseases. There are now reports which indicate that THL can inhibit growth of malignant tumors in animals. However, the mechanisms by which THL mediates its anti-tumor actions are still being explored. Because vascular endothelial growth factor-A (VEGF) induced angiogenesis plays a critical role in the pathogenesis of cancer, we therefore investigated whether tumor inhibitory effects of THL or its active constituents are through suppression of VEGF actions. We herein report that THL and chebulinic (CI) present in THL can significantly and specifically inhibit VEGF induced angiogenesis by suppressing VEGF receptor-2 (VEGFR-2) phosphorylation. These results are of clinical significance as these inexpensive and non-toxic natural products can be used for the prevention and treatment of diseases where VEGF induced angiogenesis has an important role.
Sigma-Aldrich
Chebulinic acid, ≥85% (LC/MS-ELSD)
Pricing and availability is not currently available.