Merck
  • [Metabolic detoxification of bakuchiol is mediated by cytochrome P450 enzymes in human liver microsomes].
[Metabolic detoxification of bakuchiol is mediated by cytochrome P450 enzymes in human liver microsomes].
Beijing da xue xue bao. Yi xue ban = Journal of Peking University. Health sciences (2012-06-14)
Ai-fang Li, Guo-lin Shen, Shi-yong Jiao, Hua Li, Qi Wang
ABSTRACT
To analyze cytochrome P450 (CYP) phenotyping for bakuchiol metabolism and study the mechanism of detoxification of bakuchiol by human liver microsomes (HLM) in vitro. The CYP phenotyping for bakuchiol metabolism was determined using HLM combined with CYP specific inhibitors and recombinant human CYP isoforms. The relative activities of CYP isoforms were determined by analyzing the formation of the substrate metabolites using HPLC-MS/MS, in presence or absence of 1-aminobenzotriazole (ABT) which was CYP enzymes' broad spectrum inhibitor. The residual concentrations of bakuchiol in microsomal incubates were determined using HPLC to investigate ABT's effect on the metabolism of bakuchiol. The effects of CYP enzymes on the nephrotoxicity of bakuchiol were investigated using human kidney-2(HK-2) by MTT assay, in presence or absence of ABT. CYP1A2, CYP2C9, CYP2C19 and CYP3A4 in HLM were involved in bakuchiol metabolism, among which CYP2C19 showed the highest metabolic rate. Co-incubation with ABT (2.5 mmol/L) could inhibit more than 90% of the enzyme activities for CYP1A2, CYP2C9, CYP2C19 and CYP3A4. ABT (2.5 mmol/L) could inhibit the HLM metabolism of bakuchiol with inhibition ratio 83.24%±2.13%. When preincubated with ABT, the metabolic detoxification of bakuchiol by HLM was significantly reduced (P<0.05). The mechanism of metabolic detoxification of bakuchiol by HLM is associated with bakuchiol metabolism by CYP enzymes to form non toxic or lower toxic metabolites. The broad spectrum inhibitor of CYP could inverse the detoxification of HLM.